missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Wang resin, 1% cross-linked, 0.8-1.1mmol/g, 200-400 mesh

Manufacturer:  Alfa Aesar™ L1702806

 View more versions of this product

Catalog No. AAL1702806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Wang resin
201058-08-4
5g
MFCD00131778
NERFNHBZJXXFGY-UHFFFAOYSA-N
[4-[(4-methylphenyl)methoxy]phenyl]methanol
2734007
1% cross-linked
0.8 to 1.1mmol/g
C15H16O2
4-Benzyloxybenzyl alcohol, polymer supported; Polystyrene PHB
CC1=CC=C(C=C1)COC2=CC=C(C=C2)CO
228.291

Chemical Identifiers

201058-08-4
228.291
NERFNHBZJXXFGY-UHFFFAOYSA-N
2734007
CC1=CC=C(C=C1)COC2=CC=C(C=C2)CO
C15H16O2
MFCD00131778
4-Benzyloxybenzyl alcohol, polymer supported; Polystyrene PHB
[4-[(4-methylphenyl)methoxy]phenyl]methanol
Documents