Please login to your online account to display your discounted pricing

Walter Stern Perforated Glass Beads


Spread boiling area

Manufacturer: walter stern inc  101A4MM

 View more versions of this product

Catalog No. 11-311A

  • / Pack
 

Description & Specifications

Specifications

Quantity 454g Bottle
Material Soda Lime glass
Diameter (Metric) 4mm