Please login to your online account to display your discounted pricing

Walch Top Shelf


Manufacturer: fisher science education  046275

 View more versions of this product

Catalog No. S67960

  • / Each
 

Description & Specifications

Specifications

Grade 9 to 12
Format Book
Category Forensic Science
Product Type Top Shelf Science: Forensics
For Use With (Application) Stimulates scientific inquiry through hands on lab experiments