Please login to your online account to display your discounted pricing
Wheaton™ High Performance Electronic Crimping Tool
Designed with inter-changeable jaws, so that you only need one tool and purchase the size crimper and decappers needed for your laboratory.
$2,717.87 - $4,561.04
Specifications
For Use With (Application) Crimping and decapping autosampler vials
Product Type High Performance Electronic Crimping Tool
View More Specs
Products
Catalog Number Mfr. No. Vial Closure Size Price Quantity    
02-915-010
wheaton™
W225730A*
---- Case for $2,717.87
02-915-011
wheaton™
W225731A
20mm Case for $3,563.43
02-915-012
wheaton™
W225732A
20mm Case for $4,561.04
02-915-013
wheaton™
W225733A
20mm Case for $4,561.04
Description & Specifications
Specifications
For Use With (Application) Crimping and decapping autosampler vials
Product Type High Performance Electronic Crimping Tool
  • Jaw sets are available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps
  • Able to store up to 9 separate programs for different caps and cap sizes
  • Designed with external power source and cord (no battery)
  • Ergonomically design reduces strain and arm injury associated with many manual crimping tools