W.S. TYLER™ RO-TAP™ Sieving Lab Kits
All-in-one sieving lab kits make particle analysis testing easy and convenient
Specifications
Includes One Ro-Tap Sieve Shaker, Sound Enclosure and Test Stand
View More Specs
Includes: One Ro-Tap Sieve Shaker, Sound Enclosure (04-909-5) and Test Stand (22-043-895).
Requires: Sieves and receiver (order separately)
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
04-909-1 W S Tyler Inc
RX29LABKIT
8 in. Ro-Tap RX-29 sieve shaker N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
04-909-1A W S Tyler Inc
RX30LABKIT
12 in. RX-30 Ro-Tap Wet sieve shaker N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
Ro-Tap Sieve Shaker
  • Kit for 8 in. diameter sieves holds 6 sieves (full-height and pan) or 13 sieves (half-height and pan) for particle testing, 2 in. (5cm) down through 635 mesh
  • Kit for12 in. diameter sieves hold 4 sieves (full-height and pan) or 10 sieves (half-height and pan) for testing down through 500 mesh
Order Info
Choose kit based on sieve diameter: 8 or 12 in. (20 or 30cm); for 110V; 220V shaker kits are also available through your Fisher Scientific Customer Service Representative
Description & Specifications
Specifications
One Ro-Tap Sieve Shaker, Sound Enclosure and Test Stand