Please login to your online account to display your discounted pricing

W-9 hydrochloride, Tocris

Calmodulin antagonist

$172.26 - $726.06

Chemical Identifiers

Chemical Name or Material W-9 hydrochloride
Assay Percent Range >98%
Molecular Formula C16H21ClN2O2S·HCl
Formula Weight 377.33
Synonym N-(6-Aminohexyl)-5-chloro-2-naphthalenesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

037010

 
r & d systems
RDS037010
10mg Each for $172.26

037050

 
r & d systems
RDS037050
50mg Each for $726.06
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 377.33
CAS 69762-85-2
Molecular Formula C16H21ClN2O2S·HCl
Chemical Name or Material W-9 hydrochloride
Assay Percent Range >98%
Synonym N-(6-Aminohexyl)-5-chloro-2-naphthalenesulfonamide hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature