Please login to your online account to display your discounted pricing

W-9 hydrochloride, Tocris


Calmodulin antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS037010

 View more versions of this product

Catalog No. 037010

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 377.33
CAS 69762-85-2
Molecular Formula C16H21ClN2O2S·HCl
Chemical Name or Material W-9 hydrochloride
Assay Percent Range >98%
Synonym N-(6-Aminohexyl)-5-chloro-2-naphthalenesulfonamide hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material W-9 hydrochloride
Assay Percent Range >98%
Molecular Formula C16H21ClN2O2S·HCl
Formula Weight 377.33
Synonym N-(6-Aminohexyl)-5-chloro-2-naphthalenesulfonamide hydrochloride
Accessories

Store at Room Temperature