Please login to your online account to display your discounted pricing

W-5 hydrochloride, Tocris

Calmodulin antagonist

$177.37 - $177.37

Chemical Identifiers

Chemical Name or Material W-5 hydrochloride
Assay Percent Range >98%
Molecular Formula C16H22N2O2S·HCl
Formula Weight 342.88
Synonym N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

036810

 
r & d systems
RDS036810
10mg Each for $177.37
Description & Specifications

Specifications

Chemical Name or Material W-5 hydrochloride
CAS 61714-25-8
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C16H22N2O2S·HCl
Formula Weight 342.88
Quantity 10mg
Synonym N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride
Safety and Handling

Store at Room Temperature