Please login to your online account to display your discounted pricing

W-13 hydrochloride, Tocris

Calmodulin antagonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material W-13 hydrochloride
Assay Percent Range >98%
Molecular Formula C14H17ClN2O2S·HCl
Formula Weight 349.27
Synonym N-(4-Aminobutyl)-5-chloro-2-naphthalenesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

036150

 
r & d systems
RDS036150
50mg Each for $762.58

036110

 
r & d systems
RDS036110
10mg Each for $181.19
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 349.27
Chemical Name or Material W-13 hydrochloride
Assay Percent Range >98%
Molecular Formula C14H17ClN2O2S·HCl
CAS 88519-57-7
Synonym N-(4-Aminobutyl)-5-chloro-2-naphthalenesulfonamide hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature