Please login to your online account to display your discounted pricing

VX 745, Tocris™

Potent and selective p38α inhibitor

$264.88 - $1,099.17

Chemical Identifiers

Chemical Name or Material VX 745
Assay Percent Range >98%
Molecular Formula C19H9Cl2F2N3OS
Formula Weight 436.26
Synonym 5-(2,6-Dichlorophenyl)-2-[2,4-difluorophenyl)thio]-6H-pyrimido[1,6-b]pyridazin-6-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

391510

 
r & d systems
391510
10mg Each for $264.88

391550

 
r & d systems
391550
50mg Each for $1,099.17
Description & Specifications

Specifications

Formula Weight 436.26
Quantity 10mg
Chemical Name or Material VX 745
Synonym 5-(2,6-Dichlorophenyl)-2-[2,4-difluorophenyl)thio]-6H-pyrimido[1,6-b]pyridazin-6-one
Assay Percent Range >98%
Molecular Formula C19H9Cl2F2N3OS
CAS 209410-46-8
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C