Please login to your online account to display your discounted pricing

VULM 1457, Tocris

Acyl-CoA: cholesterol acyltransferase (ACAT) inhibitor

$214.12 - $893.46

Chemical Identifiers

Chemical Name or Material VULM 1457
Assay Percent Range >99%
Molecular Formula C25H27N3O3S
Formula Weight 449.57
Synonym N-[2, 6-bis(1-Methylethyl)phenyl]-N'-[4-[(4-nitrophenyl)thio]phenyl]urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

298810

 
r & d systems
RDS298810
10mg Each for $214.12

298850

 
r & d systems
RDS298850
50mg Each for $893.46
Description & Specifications

Specifications

Formula Weight 449.57
Quantity 10mg
Synonym N-[2, 6-bis(1-Methylethyl)phenyl]-N'-[4-[(4-nitrophenyl)thio]phenyl]urea
Molecular Formula C25H27N3O3S
Chemical Name or Material VULM 1457
Assay Percent Range >99%
CAS 228544-65-8
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C