missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

VUF 5681 dihydrobromide, Tocris

Potent H3 receptor silent antagonist

$225.77 - $924.23

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C11H19N3·2HBr
Formula Weight 355.11
Synonym 4-[3-(1H-Imidazol-4-yl)propyl]piperidine dihydrobromide
Chemical Name or Material VUF 5681 dihydrobromide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
249310 R & D Systems
RDS249310
10mg Each for $225.77
249350 R & D Systems
RDS249350
50mg Each for $924.23
Specifications Description & Specifications

Chemical Identifiers

>98%
355.11
VUF 5681 dihydrobromide
C11H19N3·2HBr
4-[3-(1H-Imidazol-4-yl)propyl]piperidine dihydrobromide

Specifications

639089-06-8
C11H19N3·2HBr
4-[3-(1H-Imidazol-4-yl)propyl]piperidine dihydrobromide
VUF 5681 dihydrobromide
>98%
355.11
10mg

Safety and Handling

Recommended Storage : Desiccate at Room Temperature