Please login to your online account to display your discounted pricing

VUF 5574, Tocris


Potent, selective hA3 antagonist

Quantity 10mg

Manufacturer: r & d systems  135910

 View more versions of this product

Catalog No. 135910

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 371.39
Quantity 10mg
Chemical Name or Material VUF 5574
Assay Percent Range >98%
Molecular Formula C21H17N5O2
CAS 280570-45-8
Synonym N-(2-Methoxyphenyl)-N'-[2-(3-pyrindinyl)-4-quinazolinyl]-urea
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material VUF 5574
Assay Percent Range >98%
Molecular Formula C21H17N5O2
Formula Weight 371.39
Synonym N-(2-Methoxyphenyl)-N'-[2-(3-pyrindinyl)-4-quinazolinyl]-urea
Accessories

Store at Room Temperature