Please login to your online account to display your discounted pricing

VUF 5574, Tocris

Potent, selective hA3 antagonist

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material VUF 5574
Assay Percent Range >98%
Molecular Formula C21H17N5O2
Formula Weight 371.39
Synonym N-(2-Methoxyphenyl)-N'-[2-(3-pyrindinyl)-4-quinazolinyl]-urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

135910

 
r & d systems
135910
10mg Each for $192.67

135950

 
r & d systems
135950
50mg Each for $812.12
Description & Specifications

Specifications

Formula Weight 371.39
Quantity 10mg
Chemical Name or Material VUF 5574
Assay Percent Range >98%
Molecular Formula C21H17N5O2
CAS 280570-45-8
Synonym N-(2-Methoxyphenyl)-N'-[2-(3-pyrindinyl)-4-quinazolinyl]-urea
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature