Please login to your online account to display your discounted pricing

VUF 10460, Tocris™

Selective H4 agonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material VUF 10460
Assay Percent Range >98%
Molecular Formula C15H19N5
Formula Weight 269.34
Synonym 4-(4-Methyl-1-piperazinyl)-6-phenyl-2-Pyrimidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

476950

 
r & d systems
476950
50mg Each for $976.15

476910

 
r & d systems
476910
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 269.34
Quantity 50mg
Molecular Formula C15H19N5
Recommended Storage Store at -20°C
Synonym 4-(4-Methyl-1-piperazinyl)-6-phenyl-2-Pyrimidinamine
CAS 1028327-66-3
Chemical Name or Material VUF 10460
Assay Percent Range >98%
Safety and Handling

Store at -20°C