Please login to your online account to display your discounted pricing

VUF 10166, Tocris™

High affinity 5-HT3 receptor antagonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material VUF 10166
Assay Percent Range >98%
Molecular Formula C13H15ClN4
Formula Weight 262.74
Synonym 2-Chloro-3-(4-methyl-1-piperazinyl)quinoxaline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

453210

 
r & d systems
453210
10mg Each for $205.70

453250

 
r & d systems
453250
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 262.74
Quantity 10mg
Chemical Name or Material VUF 10166
Assay Percent Range >98%
Molecular Formula C13H15ClN4
CAS 155584-74-0
Synonym 2-Chloro-3-(4-methyl-1-piperazinyl)quinoxaline
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature