Please login to your online account to display your discounted pricing

VU 591 hydrochloride, Tocris™

Selective Kir 1.1 (ROMK) inhibitor

$199.70 - $834.54

Chemical Identifiers

Chemical Name or Material VU 591 hydrochloride
Assay Percent Range >99%
Molecular Formula C16H12N6O5
Formula Weight 368.3
Synonym 6-Nitro-2-[(6-nitro-1H-benzimidazol-2-yl)methoxymethyl]-1H-benzimidazole hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

419710

 
r & d systems
419710
10mg Each for $199.70

419750

 
r & d systems
419750
50mg Each for $834.54
Description & Specifications

Specifications

Formula Weight 368.3
Quantity 10mg
Chemical Name or Material VU 591 hydrochloride
Assay Percent Range >99%
Molecular Formula C16H12N6O5
CAS 1315380-70-1
Synonym 6-Nitro-2-[(6-nitro-1H-benzimidazol-2-yl)methoxymethyl]-1H-benzimidazole hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature