Please login to your online account to display your discounted pricing

VU 29, Tocris™

Potent mGlu5 allosteric potentiator

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material VU 29
Assay Percent Range >99%
Molecular Formula C22H16N4O3
Formula Weight 384.39
Synonym N-(1, 3-Diphenyl-1H-pyrazolo-5-yl)-4-nitrobenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

445850

 
r & d systems
445850
50mg Each for $997.23

445810

 
r & d systems
445810
10mg Each for $244.81
Description & Specifications

Specifications

Formula Weight 384.39
Quantity 50mg
Chemical Name or Material VU 29
Assay Percent Range >99%
Molecular Formula C22H16N4O3
CAS 890764-36-0
Synonym N-(1, 3-Diphenyl-1H-pyrazolo-5-yl)-4-nitrobenzamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature