Please login to your online account to display your discounted pricing

VU 0364770, Tocris™

Positive allosteric modulator at mGlu4

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material VU 0364770
Assay Percent Range >99%
Molecular Formula C12H9ClN2O
Formula Weight 232.67
Synonym N-(3-Chlorophenyl)-2-pyridinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

446910

 
r & d systems
RDS446910
10mg Each for $205.70

446950

 
r & d systems
RDS446950
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 232.67
Quantity 10mg
Chemical Name or Material VU 0364770
Assay Percent Range >99%
Molecular Formula C12H9ClN2O
CAS 61350-00-3
Synonym N-(3-Chlorophenyl)-2-pyridinecarboxamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature