Please login to your online account to display your discounted pricing

VU 0364439, Tocris™

Positive allosteric modulator at mGlu4

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material VU 0364439
Assay Percent Range >99%
Molecular Formula C18H13Cl2N3O3S
Formula Weight 422.29
Synonym N-[3-Chloro-4-[[(2-chlorophenyl)amino]sulfonyl]phenyl]-2-pyridinecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

437110

 
r & d systems
RDS437110
10mg Each for $221.01

437150

 
r & d systems
RDS437150
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 422.29
Quantity 10mg
Chemical Name or Material VU 0364439
Synonym N-[3-Chloro-4-[[(2-chlorophenyl)amino]sulfonyl]phenyl]-2-pyridinecarboxamide
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C18H13Cl2N3O3S
CAS 1246086-78-1
Safety and Handling

Store at Room Temperature