Please login to your online account to display your discounted pricing

Tocris™ VU 0361737

Selective positive allosteric modulator at mGlu4

$168.15 - $697.40

Specifications

Product Type VU 0361737
Inhibitors VU 0361737
Molecular Weight 262.69
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

370710R

 
tocris™
RDS370710
10mg Each for $168.15

370750R

 
tocris™
RDS370750
50mg Each for $697.40
Description & Specifications

Specifications

Product Type VU 0361737
Inhibitors VU 0361737
Molecular Weight 262.69
Purity >99

Selective positive allosteric modulator at mGlu4 receptors (EC50 values are 240 and 110nM at human and rat receptors respectively). Inactive at mGlu1, mGlu2, mGlu3, mGlu6 and mGlu7 receptors and displays weak activity at mGlu5 and mGlu8 receptors. Brain penetrant.

  • Chemical Name: N-(4-Chloro-3-methoxyphenyl)-2-pyridinecarboxamide
  • Formula: C13H11ClN2O2
  • CAS Number: 1161205-04-4
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99