Please login to your online account to display your discounted pricing

VU 0360223, Tocris™

Negative allosteric modulator at mGlu5

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material VU 0360223
Assay Percent Range >99%
Molecular Formula C15H9FN2S
Formula Weight 268.31
Synonym 3-Fluoro-5-(2-methyl-5-benzothiazolyl)-benzonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

439410

 
r & d systems
RDS439410
10mg Each for $221.01

439450

 
r & d systems
RDS439450
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 268.31
Quantity 10mg
CAS 1274859-33-4
Synonym 3-Fluoro-5-(2-methyl-5-benzothiazolyl)-benzonitrile
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Chemical Name or Material VU 0360223
Molecular Formula C15H9FN2S
Safety and Handling

Store at Room Temperature