Please login to your online account to display your discounted pricing

VU 0360172 hydrochloride, Tocris™

Positive allosteric modulator at mGlu5

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material VU 0360172 hydrochloride
Assay Percent Range >98%
Molecular Formula C18H15FN2O·HCl
Formula Weight 330.78
Synonym N-Cyclobutyl-6-[2-(3-fluorophenyl)ethynyl]-3-pyridinecarboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

432310

 
r & d systems
RDS432310
10mg Each for $188.60

432350

 
r & d systems
RDS432350
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 330.78
Quantity 10mg
Chemical Name or Material VU 0360172 hydrochloride
Assay Percent Range >98%
Molecular Formula C18H15FN2O·HCl
CAS 1309976-62-2
Synonym N-Cyclobutyl-6-[2-(3-fluorophenyl)ethynyl]-3-pyridinecarboxamide hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C