Please login to your online account to display your discounted pricing

VU 0357121, Tocris™

Positive allosteric modulator of mGlu5

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material VU 0357121
Assay Percent Range >99%
Molecular Formula C17H17F2NO2
Formula Weight 305.32
Synonym 4-Butoxy-N-(2, 4-difluorophenyl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

443710

 
r & d systems
RDS443710
10mg Each for $201.36

443750

 
r & d systems
RDS443750
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 305.32
Quantity 10mg
Assay Percent Range >99%
Recommended Storage Store at +4°C
Molecular Formula C17H17F2NO2
Synonym 4-Butoxy-N-(2, 4-difluorophenyl)benzamide
Chemical Name or Material VU 0357121
CAS 433967-28-3
Safety and Handling

Store at +4°C