Please login to your online account to display your discounted pricing

Tocris™ VU 0255035

Highly selective muscarinic M1 antagonist

$225.77 - $924.23

Specifications

Product Type VU 0255035
Inhibitors VU 0255035
Molecular Weight 432.52
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

372710R

 
tocris™
RDS372710
10mg Each for $225.77

372750R

 
tocris™
RDS372750
50mg Each for $924.23
Description & Specifications

Specifications

Product Type VU 0255035
Inhibitors VU 0255035
Molecular Weight 432.52
Purity >99

Highly selective muscarinic M1 antagonist (Ki = 14.87nM). Targets the M1 orthosteric site with greater than 75-fold selectivity over the other receptor subtypes. Reduces pilocarpine-induced seizures in mice. Brain penetrant.

  • Chemical Name: N-[3-Oxo-3-[4-(4-pyridinyl)-1-piperazinyl]propyl]-2,1,3-benzothiadiazole-4-sulfonamide
  • Formula: C18H20N6O3S2
  • CAS Number: 1135243-19-4
  • Solubility: Soluble to 20mM in DMSO
  • Purity: >99