Please login to your online account to display your discounted pricing

Tocris™ VU 0155069

Potent and selective PLD1 inhibitor

$264.88 - $1,099.17

Specifications

Product Type VU 0155069
Inhibitors VU 0155069
Molecular Weight 499.43
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

357510R

 
tocris™
RDS357510
10mg Each for $264.88

357550R

 
tocris™
RDS357550
50mg Each for $1,099.17
Description & Specifications

Specifications

Product Type VU 0155069
Inhibitors VU 0155069
Molecular Weight 499.43
Purity >98

Potent and selective phospholipase D1 (PLD1) inhibitor (IC50 values are 46 and 933nM for PLD1 and PLD2 respectively). Inhibits migration of human and mouse breast cancer cell lines in transwell assays.

  • Chemical Name: N-[(1S)-2-[4-(5-Chloro-2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)-1-piperidinyl]-1-methylethyl-2-naphthalenecarboxamide
  • Formula: C26H27ClN4O2.HCl
  • CAS Number: 1130067-06-9
  • Solubility: Soluble to 100mM in DMSO and to 50mM in ethanol
  • Purity: >98