Please login to your online account to display your discounted pricing

VU 0155041, Tocris

Potent, positive allosteric mGlu4 agonist

$76.90 - $330.06

Chemical Identifiers

Chemical Name or Material VU 0155041
Assay Percent Range >99%
Molecular Formula C14H15Cl2NO3
Formula Weight 316.18
Synonym cis-2-[[(3, 5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

324810

 
r & d systems
RDS324810
10mg Each for $76.90

324850

 
r & d systems
RDS324850
50mg Each for $330.06
Description & Specifications

Specifications

Formula Weight 316.18
Quantity 10mg
Chemical Name or Material VU 0155041
Assay Percent Range >99%
Molecular Formula C14H15Cl2NO3
CAS 1093757-42-6
Synonym cis-2-[[(3, 5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature