Please login to your online account to display your discounted pricing

VU 0155041 sodium salt, Tocris

Potent, positive allosteric mGlu4 agonist; Sodium salt of VU 0155041

$88.05 - $316.20

Chemical Identifiers

Chemical Name or Material VU 0155041 sodium salt
Assay Percent Range >98%
Molecular Formula C14H14Cl2NNaO3
Formula Weight 338.16
Synonym cis-2-[[(3, 5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

331150

 
r & d systems
RDS331150
50mg Each for $316.20

331110

 
r & d systems
RDS331110
10mg Each for $88.05
Description & Specifications

Specifications

Formula Weight 338.16
Quantity 50mg
Molecular Formula C14H14Cl2NNaO3
CAS 1259372-69-4
Synonym cis-2-[[(3, 5-Dichlorophenyl)amino]carbonyl]cyclohexanecarboxylic acid sodium salt
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material VU 0155041 sodium salt
Assay Percent Range >98%
Safety and Handling

Desiccate at Room Temperature