Please login to your online account to display your discounted pricing

Novus Biologicals™ VPR-66, ROR gamma T and ROR alpha Inhibitor (ROR gamma T, ROR alpha Inhibitor)


For use in research applications

Manufacturer: novus biologicals™  NBP229335

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229335

  • / Each
 

Description & Specifications

Specifications

Quantity 1mg
Inhibitors ROR gamma T, ROR alpha Inhibitor
Product Type VPR-66, ROR gamma T and ROR alpha Inhibitor
Molecular Weight 287.4
Format Crystalline, off-white solid
Storage Requirements Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.