missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Novus Biologicals™ VPR-66, ROR gamma T and ROR alpha Inhibitor (ROR gamma T, ROR alpha Inhibitor)

For use in research applications

$149.00 - $442.22

Specifications

Product Type VPR-66, ROR gamma T and ROR alpha Inhibitor
Inhibitors ROR gamma T, ROR alpha Inhibitor
Molecular Weight 287.4
Storage Requirements Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Format Crystalline, off-white solid
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP229335 Novus Biologicals™
NBP229335
1mg Each for $149.00
NBP22933510 Novus Biologicals™
NBP22933510MG
10mg Each for $442.22
Specifications Description & Specifications

Specifications

VPR-66, ROR gamma T and ROR alpha Inhibitor
287.4
Crystalline, off-white solid
ROR gamma T, ROR alpha Inhibitor
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.