missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Voriconazole, Tocris™

Triazole antifungal agent

$192.67 - $812.12

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C16H14F3N5O
Formula Weight 349.31
Synonym (2R,3S)-2-(2,4-Difluorophenyl)-3-(5-fluoro-4-pyrimidinyl)-1-((1H)-1,2,4-triazol-1-yl)-butan-2-ol
Chemical Name or Material Voriconazole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
376010 R & D Systems
376010
10mg Each for $192.67
376050 R & D Systems
376050
50mg Each for $812.12
Specifications Description & Specifications

Chemical Identifiers

>99%
349.31
Voriconazole
C16H14F3N5O
(2R,3S)-2-(2,4-Difluorophenyl)-3-(5-fluoro-4-pyrimidinyl)-1-((1H)-1,2,4-triazol-1-yl)-butan-2-ol

Specifications

137234-62-9
C16H14F3N5O
(2R,3S)-2-(2,4-Difluorophenyl)-3-(5-fluoro-4-pyrimidinyl)-1-((1H)-1,2,4-triazol-1-yl)-butan-2-ol
Voriconazole
>99%
349.31
10mg

Safety and Handling

Recommended Storage : Store at +4°C