missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ von Hippel-Lindau (vHL) Protein Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ von Hippel-Lindau (vHL) Protein Ab-1, Mouse Monoclonal Antibody.

$276.86 - $1,617.51

Specifications

Antigen von Hippel-Lindau (vHL) Protein Ab-1
Clone Ig33
Classification Monoclonal
Cross Reactivity Human,Mouse,Human,Mouse
Format Concentrated
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
MS690B Thermo Scientific™ Lab Vision™
MS690B
200μg/mL Biotin 1mL Each for $1,617.51
MS690B0 Thermo Scientific™ Lab Vision™
MS690B0
200μg/mL Biotin 100μL Each for $370.53
MS690B1 Thermo Scientific™ Lab Vision™
MS690B1
200μg/mL Biotin 500μL Each for $1,009.14
MS690P Thermo Scientific™ Lab Vision™
MS690P
200μg/mL Unlabeled 1mL Each for $1,302.69
MS690P0 Thermo Scientific™ Lab Vision™
MS690P0
200μg/mL Unlabeled 100μL Each for $276.86
MS690P1 Thermo Scientific™ Lab Vision™
MS690P1
200μg/mL Unlabeled 500μL Each for $778.98
MS690P1ABX Thermo Scientific™ Lab Vision™
MS690P1ABX
1mg/mL Unlabeled 100μL Each for $847.61
MS690PABX Thermo Scientific™ Lab Vision™
MS690PABX
1mg/mL Unlabeled 200μL Each for $1,396.67
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ von Hippel-Lindau (vHL) Protein Ab-1, Mouse Monoclonal Antibody.

Molecular genetic studies suggest that VHL is a tumor-suppressor gene. Alterations in the von Hippel-Lindau (VHL) gene are correlated with a diverse group of neoplasms including hemangioblastoma, clear cell renal carcinoma (RCC), and pheochromocytoma. Two transcriptional elongation factors, Elongin B and C, are shown to bind in vitro and in vivo to a short, colinear region of the VHL protein (pVHL) that is frequently mutated in human tumors. It is proposed that pVHL functions in vivo as a negative regulator of transcription elongation.

Host Species: Mouse

Clone: Ig33

Isotype: IgG1/κ

Species Reactivity: Human and Mouse. Others not known.

Epitope: aa 1-54

Immunogen: A recombinant protein containing aa 1-213 of human vHL

Molecular Weight: 30kDa

Positive Control: Kidney

Cellular Localization: Cytoplasmic Ab-1 does not recognize the truncated form of VHL protein.

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

von Hippel-Lindau (vHL) Protein Ab-1
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa
Ig33
Human,Mouse,Human,Mouse
Murine
Cancer and Tumor Biology
A recombinant protein containing aa 1-213 of human VHL