missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Voglibose

Orally active alpha-glucosidase inhibitor

$218.74 - $915.10

Specifications

Product Type Voglibose
Inhibitors Voglibose
Molecular Weight 267.28
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
359610R Tocris™
RDS359610
10mg Each for $218.74
359650R Tocris™
RDS359650
50mg Each for $915.10
Specifications

Description

Orally active α-glucosidase inhibitor (IC50 values are 3.9 and 6.4nM at sucrase and maltase respectively). Increases glucagon-like peptide 1 (GLP-1) secretion and decreases food consumption in ob/ob mice, and reduces plasma concentrations of glucose, triglycerides and insulin in Wistar fatty rats. Exhibits antidiabetic and antiobesity activity in vivo.
  • Chemical Name: 3,4-Dideoxy-4-[[2-hydroxy-1-(hydroxymethyl)ethyl]amino]-2-C-(hydroxymethyl)-D-epi-inositol
  • Formula: C10H21NO7
  • CAS Number: 83480-29-9
  • Solubility: Soluble to 100mM in water and to 75mM in DMSO
Description & Specifications

Specifications

Voglibose
Voglibose
267.28