Please login to your online account to display your discounted pricing

VOC Mixture (For EPA 502.2, 524.2, 8021A, 8260)

$388.91 - $388.91

Chemical Identifiers

Chemical Name or Material VOC Mixture
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-DWM-5801

 
ultra scientific
DWM5801
Ampule 1 × 1mL Each for $388.91
Description & Specifications

Specifications

Quantity 1 × 1mL
Methods For EPA Methods 502.2, 524.2, 8021A
Chemical Name or Material VOC Mixture
Concentration or Composition (by Analyte or Components) Each component at 200μg/mL in Methanol:
Benzene
Bromobenzene
Bromochloromethane
Bromodichloromethane
BromoformBromomethane
n-Butylbenzene
sec-Butylbenzene
tert-Butylbenzene
Carbon Tetrachloride
Chlorobenzene
Chloroethane
Chloroform
Chloromethane
2-Chlorotoluene
4-Chlorotoluene
Dibromochloromethane
1,2-Dibromo-3-chloropropane
1,2-DibromoethaneDibromomethane
1,2-Dichlorobenzene1,3-Dichlorobenzene
1,4-DichlorobenzeneDichlorodifluoromethane
1,1-Dichloroethane1,2-Dichloroethane
1,1-Dichloroethene
cis-1,2-Dichloroethene
trans-1,2-Dichloroethene
1,2-Dichloropropane
1,3-Dichloropropane
2,2-Dichloropropane
1,1-Dichloropropene
cis-1,3-Dichloropropene
trans-1,3-Dichloropropene
Ethylbenzene
Hexachlorobutadiene
Isopropylbenzene
4-Isopropyltoluene
Methylene Chloride
Naphthalene
n-PropylbenzeneStyrene
1,1,1,2-Tetrachloroethane
TetrachloroetheneToluene
1,2,3-Trichlorobenzene
1,2,4-Trichlorobenzene
1,1,1-Trichloroethane
1,1,2-Trichloroethane
Trichloroethene
Trichlorofluoromethane
1,2,3-Trichloropropane
1,2,4-Trimethylbenzene
1,3,5-Trimethylbenzene
Vinyl Chloride
o-Xylene
m-Xylene
p-Xylene
Packaging Ampule