Please login to your online account to display your discounted pricing

VOC Mixture, 60 Analytes, 2000μg/mL in methanol, Ultra Scientific

$514.06 - $514.06

Chemical Identifiers

Chemical Name or Material VOC Mixture
CAS 000067-56-1
CAS Max % 84.82
CAS 000074-97-5
CAS Max % 0.25
CAS 000075-27-4
CAS Max % 0.25
CAS 000075-25-2
CAS Max % 0.25
CAS 000056-23-5
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-DWM-5881

 
ultra scientific
DWM5881
1 x 1mL Each for $514.06
Description & Specifications

Specifications

Quantity 1 x 1mL
Solvent or Matrix methanol
Analyte or Component Names methanol,bromochloromethane,bromodichloromethane
Concentration or Composition (by Analyte or Components) 2000μg/mL
CAS 000056-23-5
CAS Max % 0.25
CAS Max % 84.82
CAS Max % 0.25
CAS Max % 0.25
CAS 000067-56-1
CAS Max % 0.25
Chemical Name or Material VOC Mixture
CAS 000074-97-5
CAS 000075-25-2
CAS Max % 0.25
CAS 000067-66-3
CAS 000075-27-4