Please login to your online account to display your discounted pricing

VOC Mixture, 54 Analytes, 2000μg/mL in methanol, Ultra Scientific

$444.66 - $444.66

Chemical Identifiers

Chemical Name or Material VOC Mixture
CAS 000067-56-1
CAS Max % 86.34
CAS 000074-97-5
CAS Max % 0.25
CAS 000075-27-4
CAS Max % 0.25
CAS 000075-25-2
CAS Max % 0.25
CAS 000056-23-5
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

USDWM589N1

 
ultra scientific
DWM589N1
4 x 1mL Each for $444.66
Description & Specifications

Specifications

Quantity 4 x 1mL
Analyte or Component Names methanol,bromochloromethane,bromodichloromethane
Solvent or Matrix methanol
Chemical Name or Material VOC Mixture
CAS Max % 0.25
CAS 000074-97-5
CAS 000075-25-2
CAS Max % 0.25
CAS Max % 0.25
CAS 000067-66-3
CAS 000067-56-1
CAS Max % 86.34
CAS Max % 0.25
CAS 000056-23-5
CAS Max % 0.25
Concentration or Composition (by Analyte or Components) 2000μg/mL
CAS 000075-27-4