Please login to your online account to display your discounted pricing

VOC Mixture, 54 Analytes, 2000μg/mL in methanol, Ultra Scientific


Quantity 4 x 1mL

Manufacturer: ultra scientific  DWM589N1

 View more versions of this product

Catalog No. USDWM589N1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 4 x 1mL
Analyte or Component Names methanol,bromochloromethane,bromodichloromethane
Solvent or Matrix methanol
Chemical Name or Material VOC Mixture
CAS Max % 0.25
CAS 000074-97-5
CAS 000075-25-2
CAS Max % 0.25
CAS Max % 0.25
CAS 000067-66-3
CAS 000067-56-1
CAS Max % 86.34
CAS Max % 0.25
CAS 000056-23-5
CAS Max % 0.25
Concentration or Composition (by Analyte or Components) 2000μg/mL
CAS 000075-27-4

Specifications

Chemical Name or Material VOC Mixture
CAS 000067-56-1
CAS Max % 86.34
CAS 000074-97-5
CAS Max % 0.25
CAS 000075-27-4
CAS Max % 0.25
CAS 000075-25-2
CAS Max % 0.25
CAS 000056-23-5
CAS Max % 0.25
CAS 000067-66-3
CAS Max % 0.25