Please login to your online account to display your discounted pricing

VOC Gas Mixture, 2000μg/mL in methanol, Ultra Scientific

$167.40 - $167.40

Chemical Identifiers

Chemical Name or Material VOC Gas Mixture
CAS 000067-56-1
CAS Max % 98.48
CAS 000074-83-9
CAS Max % 0.25
CAS 000075-00-3
CAS Max % 0.25
CAS 000074-87-3
CAS Max % 0.25
CAS 000075-71-8
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-DWM-5441

 
ultra scientific
801175004752
1 x 1mL Each for $167.40
Description & Specifications

Specifications

Quantity 1 x 1mL
Analyte or Component Names methanol,bromomethane,chloroethane
Solvent or Matrix methanol
CAS Max % 98.48
CAS Max % 0.25
CAS 000075-71-8
CAS Max % 0.25
CAS 000075-69-4
CAS 000074-83-9
Chemical Name or Material VOC Gas Mixture
CAS Max % 0.25
CAS 000074-87-3
CAS Max % 0.25
CAS 000067-56-1
Concentration or Composition (by Analyte or Components) 2000μg/mL
CAS Max % 0.25
CAS 000075-00-3