Please login to your online account to display your discounted pricing
Promega™ VivoGlo™ Luciferin-β-Galactoside Substrate (6-O-β-galactopyranosyl luciferin) DFS Item
Substrate for the commonly used reporter enzyme β-galactosidase. The substrate is cleaved by β-galactosidase to form luciferin and galactose. When used in a model system expressing firefly luciferase, the luciferin is then utilized in a firefly luciferase reaction to generate light.
Manufacturer: promega™  P1061
 View more versions of this product
Catalog No. PRP1061
  • / Each
Description & Specifications
Specifications
Quantity 50mg
Storage Requirements −20°C
  • Highest Quality Substrates: Eliminate potential interference in assays due to the presence of endotoxins
  • Assured Product Integrity: Most products are packaged in amber vials with septa to ensure product integrity as well as offer ease of dilution and use for imaging experiments. Product is packaged with fine tolerances to minimize the need to weigh substrates.
  • Flexibility and Convenience: Available in multiple sizes to accommodate a variety of experimental settings