Promega™ VivoGlo™ Luciferin-β-Galactoside Substrate (6-O-β-galactopyranosyl luciferin) DFS Item
Substrate for the commonly used reporter enzyme β-galactosidase. The substrate is cleaved by β-galactosidase to form luciferin and galactose. When used in a model system expressing firefly luciferase, the luciferin is then utilized in a firefly luciferase reaction to generate light.
$1,055.02 - $1,055.02
Specifications
Storage Requirements −20°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRP1061 Promega™
P1061
50mg Each for $1,055.02
Specifications
Description
  • Highest Quality Substrates: Eliminate potential interference in assays due to the presence of endotoxins
  • Assured Product Integrity: Most products are packaged in amber vials with septa to ensure product integrity as well as offer ease of dilution and use for imaging experiments. Product is packaged with fine tolerances to minimize the need to weigh substrates.
  • Flexibility and Convenience: Available in multiple sizes to accommodate a variety of experimental settings
Description & Specifications
Specifications
−20°C