Please login to your online account to display your discounted pricing

Alfa Aesar™ Vitamin A acetate in gelatin, 500,000 I.U./g

$128.38 - $503.26

Chemical Identifiers

Chemical Name or Material Vitamin A acetate
CAS 127-47-9
Name Note in gelatin
MDL Number MFCD00019413
Beilstein 1915439
Merck Index 14,10013
Synonym Retinol acetate; Retinyl acetate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1623722

 
alfa aesar™
A1623722
Each for $128.38

AAA1623736

 
alfa aesar™
A1623736
Each for $503.26

AAA1623714

 
alfa aesar™
A1623714
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Vitamin A acetate
Name Note in gelatin
CAS 127-47-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VH6825000
TSCA TSCA
MDL Number MFCD00019413
Beilstein 1915439
Merck Index 14,10013
Synonym Retinol acetate; Retinyl acetate
Concentration 500,000 IU/g
Quantity 100g
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.