missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Visualizing Mitosis Poster

Graphic depiction of plant and animal cell division.

$16.20 - $25.00

Specifications

Classification Science
For Use With (Application) Graphic depiction of plant and animal cell division
Height (English) 23 in.
Height (Metric) 58.4cm
Includes Illustrations of each mitotic stage, along with corresponding close-up photomicrographs of an onion root tip and whitefish blastula undergoing mitosis
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Material Price Quantity  
Catalog Number Mfr. No. Material Price Quantity  
S65130 Fisher Science Education
AZ-35-1025
Paper Each for $16.20
S65131 Fisher Science Education
AZ-35-1026
Laminated Each for $25.00
Specifications

Description

  • Review and compare the steps involved in both plant and animal mitosis
  • Detailed illustrations of each mitotic stage
  • Corresponding close-up photomicrographs of an onion root tip and whitefish blastula undergoing mitosis
  • Dimensions: 35W × 23 in. H (88.9 × 58.4cm)

Description & Specifications

Specifications

Science
23 in.
Illustrations of each mitotic stage, along with corresponding close-up photomicrographs of an onion root tip and whitefish blastula undergoing mitosis
88.9cm
Graphic depiction of plant and animal cell division
58.4cm
35 in.