VistaLab Technologies™ VistaStak™ Pipet Tips
Provide five stacked layers of tips for convenient use with 8- and 12-channel pipets
$117.03 - $172.63
Specifications
For Use With (Equipment) 8 and 12-channel Pipets
Product Type Tip
Sterility Nonsterile
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Tip Style Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Tip Style Volume (Metric) Price Quantity  
22-146-355 VistaLab Technologies™
VIS40609026
Large 1250μL Pack of 576 Each for $131.27
Case of 10 Pack for $1,312.70
22-146-353 VistaLab Technologies™
VIS40609024
Micro 25μL Pack of 576 Each for $117.03
22-146-354 VistaLab Technologies™
VIS40609025
Small 250μL Pack of 960 Each for $172.63
Case of 10 Pack for $1,726.30
Specifications
Description
  • Contain three to five layers of 192 tips in a 16 × 12 layout within a footprint of just 4.5 × 6 in. (11.4 × 15.2cm)
  • Each layer securely locked to a lower layer until all tips are used and the layer is discarded
  • Available in three sizes: micro, small, and large
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
8 and 12-channel Pipets
Tip
Nonsterile