Please login to your online account to display your discounted pricing
VistaLab Technologies™ VistaRak™ Pipet Tips, Nonsterile
Especially convenient for use with 8- and 12-channel pipets
$158.45 - $213.93
Specifications
For Use With (Equipment) 8 and 12-channel Pipets
Packaging Rack
Product Type Tip
View More Specs
Products
Catalog Number Mfr. No. Volume (Metric) Price Quantity    
22-146-350
vistalab technologies™
VIS40601002
25μL Case of 960 Each for $177.37
22-146-351
vistalab technologies™
VIS40602004
250μL Case of 960 Each for $158.45
22-146-352
vistalab technologies™
VIS40603004
1250μL Case of 960 Each for $213.93
Description & Specifications
Specifications
For Use With (Equipment) 8 and 12-channel Pipets
Packaging Rack
Product Type Tip
  • Contain 192 tips in a 16 × 12 layout
  • Available in three sizes: micro, small, and large
  • Sterile, pyrogen-free, and Rnase/Dnase certified tips available separately (Cat. Nos. 22-146-356 through -362)
Packaged 192 tips/rack, 5 racks/case.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.