Please login to your online account to display your discounted pricing
VistaLab Technologies™ Pipet Tips for Adjustable- and Fixed-Volume Pipets
For use with Ovation™ adjustable (100-1000μL), fixed-volume (201-1000μL), and MLA™ Brand (201-1000μL) pipets
$110.42 - $151.03
Specifications
Packaging Bulk Pack, 1000 tips/bag
View More Specs
Products
Catalog Number Mfr. No. For Use With (Equipment) Tip Style Volume (Metric) Price Quantity    
22-146-643
vistalab technologies™
VIS40583000
Ovation adjustable volume (100 to 1000uL), fixed volume (201 to 1000uL), and MLA (201 to 1000uL) Large 1400μL Pack of 1000 Each for $151.03
Case of 10 Pack for $1,510.30
22-146-636
vistalab technologies™
VIS40582000
Ovation adjustable volume (10 to 100 and 20 to 200uL), fixed volume (20 to 200uL), and MLA (5 to 200uL) Small 300μL Pack of 1000 Each for $110.42
Case of 10 Pack for $1,104.20
22-146-630
vistalab technologies™
VIS40581000
Ovation adjustable volume (0.2 to 2uL, 1.0 to 10uL, and 2.0 to 20uL) and fixed volume (1 to 20uL) Micro 35μL Pack of 1000 Each for $111.67
Case of 10 Pack for $1,116.70
Description & Specifications
Specifications
Packaging Bulk Pack, 1000 tips/bag
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.