missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
VistaLab™ Technologies Pipette Tips: Macro
For use with Ovation and MLA Brand Macro pipetters
$65.82 - $256.28
Specifications
Color | Clear |
---|---|
Rack Color | Blue |
For Use With (Equipment) | MLA™ MacroPipette, Ovation™ MacroPipette |
Shape | Straight |
Catalog Number | Mfr. No. | Volume (Metric) | Filtered | Packaging | Sterility | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Filtered | Packaging | Sterility | Price | Quantity | ||
22-146-302
|
VistaLab™ Technologies
40585000 |
5 mL | Non-Filtered | VistaBulk: Plastic Bag | Non-sterile |
Pack of 250 Each for $176.00
|
|
22-146-304
|
VistaLab™ Technologies
40585332 |
5 mL | Filtered | VistaTip: Individually Wrapped | Sterile |
Pack of 50 Each for $221.63
|
|
22-146-688
|
VistaLab™ Technologies
40585102 |
5 mL | Non-Filtered | VistaClear Box: Plastic Box | Sterile |
Pack of 60 Each for $88.00
Case of 10 Pack for $880.00 |
|
22-146-689
|
VistaLab™ Technologies
40585133 |
5 mL | Filtered | VistaClear Box: Plastic Box | Sterile |
Case of 60 Each for $130.38
|
|
22-146-321
|
VistaLab™ Technologies
40586000 |
10 mL | Non-Filtered | VistaBulk: Plastic Bag | Non-sterile |
Pack of 100 Each for $133.71
|
|
22-146-323
|
VistaLab™ Technologies
40586102 |
10 mL | Non-Filtered | VistaClear Box: Plastic Box | Sterile |
Pack of 35 Each for $117.06
|
|
22-146-322
|
VistaLab™ Technologies
40586100 |
10 mL | Non-Filtered | VistaClear Box: Plastic Box | Non-sterile |
Pack of 35 Each for $101.72
|
|
21-318-602
|
VistaLab™ Technologies
9048 |
5 mL | Non-Filtered | Paper Box | Non-sterile |
Pack of 100 Each for $65.82
Case of 10 Pack for $658.20 |
|
22-146-310
|
VistaLab™ Technologies
VIS9050 |
10 mL | Non-Filtered | Paper Box | Non-sterile |
Pack of 35 Each for $86.47
|
|
22-146-324
|
VistaLab™ Technologies
40586133 |
10 mL | Filtered | VistaClear Box: Plastic Box | Sterile |
Pack of 35 Each for $154.63
Case of 10 Pack for $1,546.30 |
|
22-146-325
|
VistaLab™ Technologies
40586332 |
10 mL | Filtered | VistaTip: Individually Wrapped | Sterile |
Pack of 50 Each for $256.28
|
|
Description
- 5 and 10 mL Macro tips packaged in Box, VistaClear Box, Bulk, and Individually Wrapped
- Box: 35 or 100 tips in a paper box, VistaClear Box: 35 or 50 tips in a clear plastic box, Bulk: 100 or 250 tips in a plastic bag, VistaTip: 50 tips individually wrapped
- Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
- All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
Clear | |
Blue | |
MLA™ MacroPipette, Ovation™ MacroPipette | |
Straight |