Please login to your online account to display your discounted pricing
VistaLab Technologies™ Ovation™ Fixed-Volume Pipetter: Two Stroke
Naturally follows the contours of the palm
$448.04 - $448.04
Specifications
Accuracy ±0.8%
Color Green
Volume (Metric) 50μL
Tip Style Small
View More Specs
Products
Catalog Number Mfr. No. Accuracy Color Volume (Metric) Price Quantity    
22-146-623
vistalab technologies™
VIS12570050
±0.8% Green 50μL Each for $448.04
22-146-629
vistalab technologies™
VIS12571000
±0.8% Purple 1000μL Each for $448.04
22-146-618
vistalab technologies™
VIS12570002
±1.5% Red 2μL Each for $448.04
Description & Specifications
Specifications
Accuracy ±0.8%
Color Green
Volume (Metric) 50μL
Tip Style Small
Two-stroke operation.
  • Adjustable hook can be easily positioned for a custom fit for left- or right-handed users
  • Fits comfortably in the palm of your hand
  • Energy release button stores the energy captured during the acquisition of tips and uses energy to push off tip at the gentle push of a button
  • Significantly improves postures and forces associated with pipetting
  • Low-force, form-fitting plunger allows user to manually control the speed of aspirating and dispensing with minimal effort
  • Nozzle with replaceable filter prevents contamination of pipetter
  • Stands securely on the counter before, during, and after the job — no risk of contamination
  • In-lab calibration adjusted with internal software
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.