Please login to your online account to display your discounted pricing

Alfa Aesar™ Viscosity standard, Specpure™, nominally 50cSt @ 40 and 7.3cSt @ 100

$141.30 - $141.30

Chemical Identifiers

Chemical Name or Material Viscosity standard
Name Note Nominally 50cSt at 40 and 7.3cSt at 100
Purity Grade Specpure™
MDL Number MFCD00084411
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47013AP

 
alfa aesar™
47013AP
500mL Each for $141.30
Description & Specifications

Specifications

Chemical Name or Material Viscosity standard
Quantity 500mL
TSCA TSCA
Name Note Nominally 50cSt at 40 and 7.3cSt at 100
Purity Grade Specpure™
Physical Form Liquid
MDL Number MFCD00084411
Odor Odorless