Please login to your online account to display your discounted pricing

Alfa Aesar™ Viscosity standard, Specpure™, nominally 500cSt @ 40 and 52cSt @ 100

Chemical Identifiers

Chemical Name or Material Viscosity standard
Name Note Nominally 500cSt at 40 and 52cSt at 100
Purity Grade Specpure™
MDL Number MFCD00084411
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47155AP

  SDS
alfa aesar™
47155AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
TSCA TSCA
Name Note Nominally 500cSt at 40 and 52cSt at 100
Chemical Name or Material Viscosity standard
Purity Grade Specpure™
Physical Form Liquid
MDL Number MFCD00084411