Please login to your online account to display your discounted pricing

Alfa Aesar™ Viscosity standard, Specpure™, nominally 120cSt @ 40 and 20cSt @ 100

$144.80 - $144.80

Chemical Identifiers

Chemical Name or Material Viscosity standard
Name Note Nominally 120cSt at 40 and 20cSt at 100
Purity Grade Specpure™
MDL Number MFCD00084411
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47100AP

  SDS
alfa aesar™
47100AP
500mL Each for $144.80
Description & Specifications

Specifications

Chemical Name or Material Viscosity standard
MDL Number MFCD00084411
Purity Grade Specpure™
Physical Form Liquid
Quantity 500mL
Name Note Nominally 120cSt at 40 and 20cSt at 100
TSCA TSCA